missing translation for 'onlineSavingsMsg'
Learn More

Alibendol 98.0+%, TCI America™

Catalog No. A29651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29651G 1 g
A29655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29651G Supplier TCI America Supplier No. A29651G
Only null left

Chemical Identifiers

CAS 26750-81-2
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.282
MDL Number MFCD00865165
InChI Key UMJHTFHIQDEGKB-UHFFFAOYSA-N
Synonym 5-Allyl-N-(2-hydroxyethyl)-3-methoxysalicylamide, 5-Allyl-2-hydroxy-N-(2-hydroxyethyl)-3-methoxybenzamide, Cebera
PubChem CID 71916
IUPAC Name 2-hydroxy-N-(2-hydroxyethyl)-3-methoxy-5-prop-2-enylbenzamide
SMILES COC1=C(C(=CC(=C1)CC=C)C(=O)NCCO)O

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 251.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Alibendol
RTECSNumber CU8730000
TSCA No