missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alginic Acid, NF, Spectrum™ Chemical
AL172, 9005-32-7, (C6H8O6)n
$269.39 - $20942.00
Chemical Identifiers
| CAS | 9005-32-7 |
|---|---|
| Molecular Formula | (C6H8O7)A(C6H8O7)B |
| SMILES | O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600390
|
Spectrum Chemical Mfg Cor
AL172125GM |
125 g | Poly Bottle |
Each for $269.39
|
|
||||
|
18600391
|
Spectrum Chemical Mfg Cor
AL172500GM |
500 g | Poly Bottle |
Each for $901.58
|
|
||||
|
18600392
|
Spectrum Chemical Mfg Cor
AL17225KG |
2.5 kg | Poly Pail |
Each for $2,924.39
|
|
||||
|
18600394
|
Spectrum Chemical Mfg Cor
AL17220KGBL |
20 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18600393
|
Spectrum Chemical Mfg Cor
AL17225KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Alginic Acid, NF is used as a tablet disintigrant. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 9005-32-7 | |
| (C6H8O7)A(C6H8O7)B | |
| O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O |
Specifications
| 9005-32-7 | |
| 15% | |
| (C6H8O7)A(C6H8O7)B | |
| O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O |
| 100% | |
| Poly Bottle | |
| 125 g | |
| NF |