missing translation for 'onlineSavingsMsg'
Learn More

Alginic Acid, NF, Spectrum™ Chemical

Catalog No. 18600390
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
20 kg
25 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600390 125 g Poly Bottle
18600391 500 g Poly Bottle
18600392 2.5 kg Poly Bottle
18600394 20 kg Poly Bottle
18600393 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600390 Supplier Spectrum Chemical Mfg Cor Supplier No. AL172125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

AL172, 9005-32-7, (C6H8O6)n

Spectrum™ Chemical Alginic Acid, NF is used as a tablet disintigrant. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-32-7
Molecular Formula (C6H8O7)A(C6H8O7)B
SMILES O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O

Specifications

CAS Max % 100%
Loss on Drying 15%
Packaging Poly Bottle
Quantity 125 g
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Alginic Acid