missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alginic acid

CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B

$114.08 - $2209.08

Chemical Identifiers

CAS 9005-32-7
Molecular Formula (C6H8O7)A(C6H8O7)B
MDL Number MFCD00081309
Synonym 2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC270581000
SDS View Documents
Thermo Scientific Chemicals
270581000
100 g Plastic bottle
Each for $114.08
Only null left
 
AC270585000
SDS View Documents
Thermo Scientific Chemicals
270585000
500 g Plastic bottle
Each for $432.59
Only null left
 
AC270580025
SDS View Documents
Thermo Scientific Chemicals
270580025
2.5 kg Plastic bottle
Each for $2,209.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9005-32-7
(C6H8O7)A(C6H8O7)B
MFCD00081309
2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid

Specifications

9005-32-7
Beige to White
15% max. (2 g, 105°C, 4 hrs)
Plastic bottle
MFCD00081309
15,235
Solubility in water: insoluble. Other solubilities: slightly soluble in hot water;,insoluble in organic solvents;,dissolves slowly in solutions of sodium carbonate,,sodium hydroxide and trisodium phosphate
Powder
>300°C
4% max.
Authentic
(C6H8O7)A(C6H8O7)B
100 g
2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid
19 to 25% (carboxyl groups) (on dried substance)
Alginic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-680-1

RTECSNumber : AZ5775000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only