missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alginic acid

CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B

Supplier:  Thermo Scientific Chemicals 270581000

 View more versions of this product

Catalog No. AC270581000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Alginic acid
>300°C
4% max.
Authentic
(C6H8O7)A(C6H8O7)B
100 g
2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid
19 to 25% (carboxyl groups) (on dried substance)
9005-32-7
Beige to White
15% max. (2 g, 105°C, 4 hrs)
Plastic bottle
MFCD00081309
15,235
Solubility in water: insoluble. Other solubilities: slightly soluble in hot water;,insoluble in organic solvents;,dissolves slowly in solutions of sodium carbonate,,sodium hydroxide and trisodium phosphate
Powder

Chemical Identifiers

9005-32-7
(C6H8O7)A(C6H8O7)B
MFCD00081309
2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-680-1

RTECSNumber : AZ5775000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only