missing translation for 'onlineSavingsMsg'
Learn More

Alfuzosin hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015611
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015611 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015611 Supplier Merck Emd Millipore Supplier No. PHR16381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

Packaging Ampule
CAS 81403-68-1
Linear Formula C19H27N5O4 · HCl
MDL Number MFCD00879135
Synonym N-(3-((4-Amino-6,7-dimethoxy-2-quinazolinyl)methylamino)propyl)tetrahydro-2-furancarboxamide hydrochloride; Uroxatral hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 425.91
Grade Certified Reference Material