missing translation for 'onlineSavingsMsg'
Learn More

Alfentanil hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. A0711ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0711ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0711ML Supplier MilliporeSigma Supplier No. A0711ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • A Snap-N-Spike™ Reference Solution suitable as starting material for use in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing and pharmaceutical research.
  • Alfentanil is a pharmaceutical synthetic opioid sold as Alfenta and Rapifen for pain relief and anesthesia.

Specifications

Chemical Name or Material Alfentanil HCl
Packaging Ampule
CAS 69049-06-5
Molecular Formula C21H33ClN6O3
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 452.98
Quantity 1 mL
Grade Certified
Show More Show Less