missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfacalcidol 95.0+%, TCI America™
$195.73 - $697.16
Chemical Identifiers
| CAS | 41294-56-8 |
|---|---|
| Molecular Formula | C27H44O2 |
| Molecular Weight (g/mol) | 400.647 |
| MDL Number | MFCD00868328 |
| InChI Key | OFHCOWSQAMBJIW-AQVMIQFASA-N |
| Synonym | alphacalcidol |
| PubChem CID | 87091364 |
| IUPAC Name | (1R,3R,5Z)-5-[2-[(1R,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol |
| SMILES | CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CC(C3=C)O)O)C |
Chemical Identifiers
| 41294-56-8 | |
| 400.647 | |
| OFHCOWSQAMBJIW-AQVMIQFASA-N | |
| 87091364 | |
| CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CC(C3=C)O)O)C |
| C27H44O2 | |
| MFCD00868328 | |
| alphacalcidol | |
| (1R,3R,5Z)-5-[2-[(1R,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol |
Specifications
| 41294-56-8 | |
| White-Yellow | |
| MFCD00868328 | |
| 2811 | |
| OFHCOWSQAMBJIW-AQVMIQFASA-N | |
| (1R,3R,5Z)-5-[2-[(1R,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol | |
| 87091364 | |
| ≥95.0% (HPLC) | |
| Alfacalcidol |
| 136°C | |
| C27H44O2 | |
| 10 mg | |
| alphacalcidol | |
| CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CC(C3=C)O)O)C | |
| 400.647 | |
| 400.65 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer (-20 °C)