CAS RN 41294-56-8
CAS RN 41294-56-8
IUPAC Name:
(1R,3S,5Z)-5-{2-[(1R,3aS,4E,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
Synonyms:
etα oksidevit gry brand of alfacalcidol hydroxycholecalciferol 1α-OHD3 alfacalcidol alfacalcidol (product) 1-hydroxycholecalciferol teva brand of alfacalcidol aps brand of alfacalcidol
Molecular Weight (g/mol):
400.65
Molecular Formula:
C27H44O2
InChi Key:
OFHCOWSQAMBJIW-AVJTYSNKSA-N
SMILES:
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)C[C@H](O)C1=C
1
–
1
of
1
results
1
Alfacalcidol 95.0+%, TCI America™
CAS: 41294-56-8 Molecular Formula: C27H44O2 Molecular Weight (g/mol): 400.647 MDL Number: MFCD00868328 InChI Key: OFHCOWSQAMBJIW-AQVMIQFASA-N Synonym: alphacalcidol PubChem CID: 87091364 IUPAC Name: (1R,3R,5Z)-5-[2-[(1R,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol SMILES: CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CC(C3=C)O)O)C