missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 647 Hydroxylamine

Catalog No. a30632
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30632 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30632 Supplier Invitrogen™ Supplier No. A30632

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor™ 647 Hydroxlamine is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins.

Alexa Fluor™ 647 Hydroxlamine is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins. Alexa Fluor™ 647 is a bright, far red fluorescent dye with excitation ideally suited to the 633 nm laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 647 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 647 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

Detailed information about this AlexaFluor™ hydroxlamine:
• Fluorophore label : Alexa Fluor™ 647 dye
• Reactive group: hydroxlamine
• Reactivity: Aldehydes or ketones
• Ex/Em of the conjugate: 651/672 nm
• Extinction coefficient: 250,000 cm-1M-1
• Spectrally similar dyes: APC, Cy5
• Molecular weight: ∼1,200

Cell Tracking and Tracing Applications
Alexa Fluor™ hydrazides and hydroxlamines are useful as low molecular weight, membrane-impermeant, aldehyde-fixable cell tracers, exhibiting brighter fluorescence and greater photostability than cell tracers derived from other spectrally similar fluorophores. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent (Cat. No. I14402).

Glycoprotein and Polysaccharide Labeling Applications
The Alexa Fluor™ hydrazides and hydroxlamines are reactive molecules that can be used to add a fluorescent label to biomolecules containing aldehydes or ketones. Aldehydes and ketones can be introduced into polysaccharides and glycoproteins by periodate-mediated oxidation of vicinal diols. Galactose oxidase can also be used to oxidize terminal galactose residues of glycoproteins to aldehydes.

Hydrazide vs Hydroxylamine
Hydrazine derivatives react with ketones and aldehydes to yield relatively stable hydrazones. Hydroxylamine derivatives (aminooxy compounds) react with aldehydes and ketones to yield oximes. Oximes are superior to hydrazones with respect to hydrolytic stability. Both hydrazones and oximes can be reduced with sodium borohydride (NaBH4) to further increase the stability of the linkage.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50μg (A33087)
Antibody Conjugate Purification kit for 50-100μg (A33088)

Specifications

Quantity 1 mg
Product Type Hydroxylamine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Hydroxylamine
Label or Dye Alexa Fluor™ 647
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 672 nm
Excitation 651 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.