missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alexa Fluor™ 594 Tyramide SuperBoost™ Kit, goat anti-mouse IgG
Description
SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).
Features:
- High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
- Simple protocol and detection using standard filters
- Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
- Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
Benefits:
- Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
- Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
- Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
- Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals
Shelf Life:
- Stable for 6 months after receiving
Specifications
Specifications
| Conjugate | Alexa Fluor 594 |
| Quantity | Sufficient for 150 Slides |
| Content And Storage | 1 kit sufficient for 150 microscope slides (18 x 18 mm), containing: Blocking buffer (1X), 22.5 mL |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Product Line | SuperBoost |
| Product Type | Tyramide Kit |
For Research Use Only. Not for use in diagnostic procedures.