missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 594 Cadaverine

Catalog No. A30678
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30678 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30678 Supplier Invitrogen™ Supplier No. A30678

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor™ 594 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via a carboxylic acid moiety.

Alexa Fluor™ 594 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via a carboxylic acid moiety. Alexa Fluor™ 594 is a bright, red dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 594 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 594 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

Detailed information about this AlexaFluor™ cadaverine:
• Fluorophore label: Alexa Fluor™ 594 dye
• Reactive group: cadaverine
• Reactivity: carboxylic acids, aldehydes, and ketones (and glutamine residues through an enzyme-catalyzed transamidation reaction)
• Ex/Em of the conjugate: 588/612 nm
• Extinction coefficient: 105,000 cm-1M-1
• Spectrally similar dyes: Texas Red
• Molecular weight: 806.94

Cell Tracking and Tracing Applications
Alexa Fluor™ cadaverines make excellent fluorescent polar tracers because they are bright, small, and water soluble. Since they contain an aldehyde-fixable functional group, they can be fixed in cells by treatment with formaldehyde or glutaraldehyde. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent (Cat. No. I14402).

Protein Labeling Applications
Alexa Fluor™ cadaverines can be used as reactive molecules for adding a fluorescent label to carboxylic acids using a coupling agent such as a carbodiimide; they do not spontaneously react with carboxylic acids in solution. They do, however, react spontaneously with the common amine-reactive functional groups, including succinimidyl esters and isothiocyanates. The amine-containing Alexa Fluor™ cadaverines can also be used to label glutamine residues in some proteins and peptides via an enzyme-catalyzed transamidation reaction.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Specifications

Quantity 1 mg
Product Type Cadaverine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Cadaverine
Label or Dye Alexa Fluor™ 594
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 612 nm
Excitation 588 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.