missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 594 Azide (Alexa Fluor™ 594 Carboxamido-(6-Azidohexanyl), Triethylammonium Salt), mixed isomers

Catalog No. a10270
Encompass
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10270 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10270 Supplier Molecular Probes™ Supplier No. A10270

Alexa Fluor™ 594 Azide (Alexa Fluor™ 594 Carboxamido-(6-Azidohexanyl), Triethylammonium Salt), mixed isomers

The red-fluorescent Alexa Fluor™ 594 azide can be reacted with terminal alkynes via a copper-catalyzed click reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.

Specifications

Format Solid
Quantity 0.5 mg
Product Type Azide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccate and protect from light.
Chemical Reactivity Alkyne
Detection Method Fluorescence
Shipping Condition Room Temperature
System Type Click-iT™
Reactive Moiety Amine, Azide
Label or Dye Alexa Fluor™ 594
Label Type Alexa Fluor
Color Red, Red
Product Line Alexa Fluor, Molecular Probes
Emission 617
Excitation 590
Show More Show Less
I am observing no signal or very low specific signal for my click-labeled samples. What can I do to improve the signal?

The click reaction is only effective when copper is in the appropriate valency. Azides and alkynes will not react with each other without copper. Make sure that the click reaction mixture is used immediately after preparation when the copper (II) concentration is at its highest.
Do not use additive buffer that has turned yellow; it must be colorless to be active.
Cells need to be adequately fixed and permeabilized for the TdT enzyme and click reagents to have access to the nucleus. Tissue samples require digestion with proteinase K or other proteolytic enzymes for sufficient TdT access.
Some reagents can bind copper and reduce its effective concentration available to catalyze the click reaction. Do not include any metal chelator (e.g., EDTA, EGTA, citrate, etc.) in any buffer or reagent prior to the click reaction. Avoid buffers or reagents that include other metal ions that may be o xidized or reduced. It may be help to include extra wash steps on the cell or tissue sample before performing the click reaction.
You can repeat the click reaction with fresh reagents to try to improve signal. Increasing the click reaction time longer than 30 minutes will not improve a low signal. Performing a second, 30 minute incubation with fresh click reaction reagents is more effective at improving labeling.
Your cells may not be apoptotic. Prepare a DNase I-treated positive control to verify that the TdT enzymatic reaction and click labeling reaction are working correctly.

I am observing high non-specific background when I image my Click-iT EdU TUNEL-labeled samples. What is causing this and what can I do to reduce the background?

The click reaction is very selective between an azide and alkyne. No other side reactions are possible in a biological system. Any non-specific background is due to non-covalent binding of the dye to various cellular components. The Select FX Signal Enhancer is not effective at reducing non-specific charge-based binding of dyes following the click reaction; we do not recommend its use with the Click-iT detection reagents. The best method to reduce background is to increase the number of BSA washes. You should always do a no-dye or no-click reaction control under the same processing and detection conditions to verify that the background is actually due to the dye and not autofluorescence. You should also perform the complete click reaction on a no-TdT enzyme control sample to verify the specificity of the click reaction signal.

I notice that when I post-stain my cells with DAPI after performing the click reaction to detect EdU incorporation, my DAPI signal is lower compared to my no-click reaction control samples. What causes the reduction in DAPI signal?

The copper in the click reaction denatures DNA to a small extent (although not as much as is required for efficient BrdU detection), which can affect the binding affinity of DNA dyes including DAPI and Hoechst stain. This effect should only be apparent with the classic EdU kits and not the Click-iT Plus EdU kits, which use a lower copper concentration.

I am observing no signal or very low signal for my click-labeled samples. What can I do to improve the signal?

The click reaction is only effective when copper is in the appropriate valency. Except for the DIBO alkyne-azide reaction, azides and alkynes will not react with each other without copper. Make sure that the click reaction mixture is used immediately after preparation when the copper (II) concentration is at its highest.
Do not use additive buffer that has turned yellow; it must be colorless to be active.
Cells need to be adequately fixed and permeabilized for the click reagents to have access to intracellular components that have incorporated the click substrate(s).
Some reagents can bind copper and reduce its effective concentration available to catalyze the click reaction. Do not include any metal chelator (e.g., EDTA, EGTA, citrate, etc.) in any buffer or reagent prior to the click reaction. Avoid buffers or reagents that include other metal ions that may be oxidized or reduced. It may be help to include extra wash steps on the cell or tissue sample before performing the click reaction.
You can repeat the click reaction with fresh reagents to try to improve signal. Increasing the click reaction time longer than 30 minutes will not improve a low signal. Performing a second, 30 minute incubation with fresh click reaction reagents is more effective at improving labeling.
Low signal can also be due to low incorporation of EdU, EU, or other click substrates. Other click substrates (e.g., AHA, HPG, palmitic acid, azide, etc.) incorporated into cellular components may have been lost if not adequately cross-linked in place or if the wrong fixative was used. For click substrates that are incorporated into the membrane or lipids, you should avoid the use of alcohol or acetone fixatives and permeabilizing agents.
The incorporated click substrate must be accessible at the time of the click reaction; labeling of incorporated amino acid analogs may be lower in native proteins relative to denatured proteins.
You may need to optimize the metabolic labeling conditions including analog incubation time or concentration. Cells that are healthy, not too high of a passage number and not too crowded may incorporate the analog better. You may create a positive control by including extra doses of the click substrate during multiple time points during an incubation time that spans or closely spans the doubling time of the cell type of interest.

I am observing high background when I analyze my click-labeled samples. What is causing this and what can I do to reduce the background?

The click reaction is very selective between an azide and alkyne. No other side reactions are possible in a biological system. Any non-specific background is due to non-covalent binding of the dye to various cellular components. The Select FX Signal Enhancer is not effective at reducing non-specific charge-based binding of dyes following the click reaction; we do not recommend its use with the Click-iT detection reagents. The best method to reduce background is to increase the number of BSA washes. You should always do a no-dye or no-click reaction control under the same processing and detection conditions to verify that the background is actually due to the dye and not autofluorescence. You can also perform the complete click reaction on a carrier solvent-only, no EdU or no-EU control to verify the specificity of the click reaction signal.

Can I perform Click-iT EdU detection on live cells?

No, the EdU metabolic labeling reagent must be used on live cells, but the actual click detection reaction must be performed on fixed and permeabilized samples, as the azide detection reagents and buffer components are cell impermeant.

Can I combine Click-iT EdU labeling with EdU TUNEL labeling so that I can detect proliferation and apoptosis in the same sample?

It is possible, but if you have not completely labeled all of the metabolically incorporated EdU in the first click reaction, then it will be labeled in the second click reaction for TUNEL labeling, leading to false positives for apoptotic cells. It would be simpler to combine Click-iT EdU labeling with BrdU TUNEL labeling, as BrdU detection will not cross-react with EdU labeled cells. If you really wish to perform a double EdU labeling for both proliferation and apoptosis detection, then you should repeat the click reaction to detect the metabolically incorporated EdU using fresh click reagents to ensure that all of the incorporated EdU is labeled before performing the EdU TUNEL assay. You should then perform a control no-TdT enzyme EdU TUNEL assay to verify that there is no signal generated with the TUNEL click reaction.

Can I combine EdU and BrdU labeling and detection on the same sample?

Yes, EdU and BrdU labeling can be combined for dual-pulse labeling of cell proliferation in cultured cells and in vivo. BrdU will be preferentially incorporated into DNA, so perform the EdU incubation first followed by the BrdU incubation. Removal of EdU from the media is not required in cultured cells when BrdU is added as the second label. Perform an alcohol fixation followed by some method of DNA denaturation as required for the BrdU detection protocol and then perform the click labeling reaction for detection of EdU followed by antibody labeling for detection of BrdU. Be sure to select a BrdU antibody that does not have cross-reactivity to EdU, such as our MoBU-1 clone (Cat. No. B35141). Many BrdU antibodies have been shown to have some amount of cross-reactivity with incorporated EdU. Here is a link (http://www.thermofisher.com/us/en/home/references/protocols/cell-and-tissue-analysis/flow-cytometry-protocol/cell-proliferation/dual-pulse-labeling-of-cell-proliferation-using-edu-and-brdu-incorporation.html) to an example protocol for dual-pulse labeling using EdU and BrdU.

Can I perform Click-iT EdU detection on cells growing in 3D culture?

We have not validated the use of EdU for proliferation in 3D culture systems, but as this reagent is compatible for labeling cells in vivo, it is also expected to label cells in 3D culture systems. There are a number of reports in the literature that use this product in 3D culture systems; here are some citations:

Lei Y, Schaffer DV (2013) A fully defined and scalable 3D culture system for human pluripotent stem cell expansion and differentiation. Proc Natl Acad Sci U S A 110:E5039-E5048.
Derda R, Laromaine A, Mammoto A et al. (2009) Paper-supported 3D cell culture for tissue-based bioassays. Proc Natl Acad Sci U S A 106:18457-18462.
Robertson FM, Ogasawara MA, Ye Z et al. (2010) Imaging and Analysis of 3D Tumor Spheroids Enriched for a Cancer Stem Cell Phenotype. J Biomol Screen 15:820-829.

Can I use EdU in vivo and do you have a recommended protocol?

Yes, EdU is frequently used for in vivo assays and acceptable EdU incorporation can be obtained following injection or media incubation. Optimal EdU incubation times and concentrations will depend on the incorporation method and the organism/tissue type. Recommendations on EdU incorporation methods in various model organisms and protocols for detection of EdU in tissue samples can be found in the In Vivo Use of Click-iT EdU Cell Proliferation Assays application note and the Click-iT EdU Labeling In Vivo Cell Proliferation Protocol.

Table 2 of the EdU (5-ethynyl-2'-deoxyuridine) product manual also provides some recommendations. Methods that have been optimized for BrdU labeling for your organism are a good starting point. Tissue samples can be frozen or paraffin-embedded and then processed using normal procedures for subsequent IHC staining. For detection of EdU, use any desired method for fixation and permeabilization, and then perform the click detection reaction exactly according to the protocol for cultured cells in the product manual.

I am collecting samples over time and would like to perform the Click-iT detection reaction on all the samples at the same time. Are there stopping points in the protocol, so that I do not have to perform the entire detection procedure in the same day?

Yes, you can store samples after fixing in formaldehyde and washing, before the permeabilization step. Just keep the cells in PBS, cover and seal the container well, and store at 4 degrees C. The cells should be fine for at least a week. You can also store the samples after the click reaction and wash steps and then perform any immunostaining and nuclear counterstaining on the following day.

I want to analyze my EdU labeled cells by flow and imaging. Do I have to purchase separate EdU flow and imaging kits or can I just use one kit for both assays?

The Click-iT EdU flow kits use a lower azide dye concentration than what is optimal for the EdU imaging kits, so it is not possible to use a flow kit to label cells for an imaging experiment. It is possible to use a Click-iT EdU imaging kit for samples for flow analysis, but it would be necessary to optimize the azide dye concentration first. The azide dye concentrations used in the Click-iT EdU kits are proprietary.

I ran out of the copper sulfate solution included in the Click-iT EdU, RNA and buffer kits. What can I use?

The copper sulfate solution component supplied in the Click-iT kits is 100 mM copper sulfate in water. Copper (II) sulfate (CuSO4) powder is available from many chemical suppliers.

If I run out of the azide dye stock provided in the Click-iT assay kit or I would like to substitute a different color, what concentration of the stand-alone reagent should I use?

Unfortunately, the actual concentration of the azide dyes used in the Click-iT assay kits is proprietary, so we cannot provide the exact concentration used. For a general guideline, we recommend making up a 1-10 mM DMSO stock solution and using at approximately 1-10 µM final concentration in the click reaction. Too high of a dye concentration can result in high nonspecific background or dye aggregates; too low of a concentration may not give a strong enough signal.

I am metabolically incorporating an alkyne-containing reagent into my cells. What do I need to purchase to perform the Click-iT reaction?

If you are using EdU or EU, the simplest thing to do is purchase a complete kit that is specific for the type of assay you are performing, either flow, imaging, or a microplate assay. The complete kits contain the azide detection reagent as well as copper and the buffers necessary to perform the click reaction. If you have a different alkyne metabolite analog or do not wish to purchase EdU or EU, then you will need to purchase any of our azide dye detection reagents and the Click-iT Cell Reaction Buffer Kit (Cat. No. C10269), which contains the copper solution and the other buffer reagents necessary to perform the click reaction for flow and imaging assays. If you wish to perform a microplate assay, then you are limited to the Click-iT EdU Microplate Assay (Cat. No. C10214), which provides all the reagents necessary for the click reaction as well as the reagents necessary to perform the amplification of the signal so that it can be detected on a microplate reader.


For Research Use Only. Not for use in diagnostic procedures.