missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 568 NHS Ester (Succinimidyl Ester)

Catalog No. A20003
Encompass
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20003 1 mg
A20103 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20003 Supplier Invitrogen™ Supplier No. A20003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor™ 568 is a bright orange dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 568 dye is water soluble and pH-insensitive from pH 4 to pH 10.

Alexa Fluor™ 568 is a bright orange dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 568 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 568 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 568 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:
Fluorophore label: Alexa Fluor™ 568 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 578/602 nm
Extinction coefficient: 88,000 cm-1M-1
Spectrally similar dyes: Rhodamine Red
Molecular weight: 791.8

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (Cat. No. A33086)
Antibody Conjugate Purification Kit for 20-50 μg (Cat. No. A33087)
Antibody Conjugate Purification kit for 50-100 μg (Cat. No. A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Specifications

Quantity 1 mg
Product Type Dye
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Alexa Fluor™ 568
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 602 nm
Excitation 578 nm
Show More Show Less
I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

For Research Use Only. Not for use in diagnostic procedures.