missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 555 Tyramide SuperBoost™ Kit, streptavidin

Catalog No. B40933
Encompass
Change view
Click to view available options
Quantity:
150 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40933 150 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40933 Supplier Invitrogen™ Supplier No. B40933

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SuperBoost™ tyramide signal amplification is the most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).

SuperBoost kits combine the brightness of AlexaFluor™ dyes with the superior signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce a sensitivity 10-200 times greater than standard methods. For standout research, SuperBoost kits sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal.

SuperBoost kits are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a SuperBoost kit can be imaged using any type of microscope, producing high-resolution multiplex images. This particular kit features AlexaFluor 555 tyramide (555/565 ex/em), detected using a standard Orange/RFP/TRITC filter cube. This kit also features HRP-conjugated streptavidin.

• Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
• Simple protocol and detection using standard filters
• Suitable for high-resolution multiplex images—co-label with DAPI, secondary antibodies, and other SuperBoost kits
• Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments

SuperBoost kits are based on the tyramide signal amplification system, which uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labeling of a target protein or nucleic acid sequence in situ. A typical ICC/IHC/ISH experiment using a SuperBoost kit requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments. SuperBoost kits offer superior specific signal intensity over background, so the protocol is easily optimized to detect specific signal in samples where high endogenous autofluorescence is observed.

Enhancement of signal using Alexa Fluor tyramides: SuperBoost kits utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules. SuperBoost kits are the only kits that combine the brightness of Alexa Fluor dyes with the enhancement of tyramide signal amplification to produce a superior signal.

Reaction stop solution: Like any enzyme-based labeling system, it is possible to overdevelop the signal. SuperBoost kits include an HRP stop solution to halt the HRP reaction. HRP stop solution can be used to obtain maximum signal, without increase of background signal. Images produced with optimized HRP reaction times are as sharp as images produced with standard ICC/IHC/ISH methods, but with 10-200 times more sensitivity.

Reduction of background: SuperBoost kits include blockers for the elimination or reduction of endogenous peroxidase and fluorescent background signals. These blockers help ensure that only specific signals are enhanced while keeping non-specific/background signals in check.

Specifications

Conjugate Alexa Fluor 555
Quantity 150 Slides
Content And Storage 1 kit sufficient for 150 microscope slides (18 x 18 mm), containing:

• Blocking buffer (1 X), 22.5 mL
• HRP-conjugated streptavidin (1 X), 22.5 mL
• Alexa Fluor tyramide reagent
• Hydrogen peroxide (stabilized 3% solution), 28.5 mL
• Reaction buffer (20 X), 6 mL
• Reaction stop solution, 8 mg
• Dimethylsulfoxide (DMSO), 200 μL

Store (at 2°C to 8°C. When stored as directed, the product is stable for 6 months after receiving.)
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Line SuperBoost
Product Type Tyramide Kit
With a SuperBoost tyramide kit, I got excessive and non-specific labeling. What can I do to limit background and acquire a more localized labeling?

To limit background, we recommend performing a pre-blocking step with 3% H2O2 for 60 mins to inactivate endogenous peroxidases. To limit the localization of labeling, we recommend optimizing the final concentration of the primary and secondary antibodies and the dye-tyramide. You may also limit the incubation time of the dye-tyramide on the sample.

Is it possible to perform dual TSA labeling with SuperBoost tyramide kits?

Yes. This involves the sequential application of the antibodies and the tyramides with a HRP-quenching step between antibodies using H2O2.

How are SuperBoost tyramide kits different from the original TSA labeling kits?

The SuperBoost tyramide kits utilize poly-HRP labeled antibodies. This provides a greater number of horseradish peroxidase (HRP) molecules per antibody. The original kits used antibodies and streptavidin that were directly conjugated with HRP and thus, limited the number per antibody or streptavidin.

For Research Use Only. Not for use in diagnostic procedures.