missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 555 Microscale Protein Labeling Kit

Catalog No. A30007
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30007 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30007 Supplier Molecular Probes™ Supplier No. A30007

Microscale Protein Labeling Kits provide a convenient means for attaching a fluorescent label to a small amount of antibody or protein (20–100 μg).

Microscale Protein Labeling Kits provide a convenient means for attaching a fluorescent label to a small amount of antibody or protein (20–100μg). The kits are available in four Alexa Fluor™ colors (or biotin) and supply everything needed for three labeling and separation reactions.

Important Features of Microscale Protein Labeling Kits:
• Labeled proteins typically ready to use typically in 2 hours (∼30 minutes hands-on time)
• Optimized for 20–100μg of protein with molecular weights between 12 and 150 kDa
• Purified using convenient spin filters with yields between 60 and 90%
• Stabilizing proteins must be removed from the sample before labeling

Stable Reaction Chemistry and Superior Alexa Fluor™ Dyes
In the Microscale Protein Labeling Kits, the reactive dye contains a succinimidyl (NHS) ester moiety that reacts with primary amines of proteins to form stable dye-protein conjugates. Compared to traditional dyes, Alexa Fluor™ dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. And generally when using Alexa Fluor™ dyes, higher degrees of labeling can be achieved without intramolecular quenching. For details see Alexa Fluor™ Dyes Spanning the Visible and Infrared Spectrum—Section 1.3.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.

We’ll Make a Custom Antibody Conjugate for You
If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Specifications

Product Type Labeling Kit
Quantity 1 kit
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Detection Method Fluorescence
Excitation/Emission 555/565 nm
Labeling Method Conjugation-based
Labeling Scale 20–100 μg
Shipping Condition Room Temperature
Product Line Alexa Fluor
Reactive Moiety Succinimidyl (NHS) Ester
Label or Dye Alexa Fluor 555
Color Orange
Labeling Target Proteins
Label Type Alexa Fluor
Show More Show Less
Can I use 50 μg of protein with Fluorescent Protein Labeling Kits?

No. We recommend using 1 mg of protein with Fluorescent Protein Labeling Kits. For smaller protein sample sizes, we recommend using Microscale Protein Labeling kits which are optimized for 20-100 µg of protein.

What formulation of antibody should I use for conjugation for small animal in vivo imaging?

To allow for good reaction kinetics, antibodies should be in PBS buffer at a concentration of 0.5-3.0 mg/ml. The antibody must be free of preservatives (azide etc.), amine containing buffers and carrier proteins such as BSA.

What is degree of labeling (DOL)?

Degree of labeling (DOL) describes the number of fluorophores per antibody. For in vivo labeling experiments, the DOL is restricted to a narrow range because it has significant consequences for the biodistribution and clearance of the probe. For example, for in vivo imaging, we have determined that the DOL range for the far-red Alexa Fluor dyes is 1.5 to 3 molecules per antibody for optimal optical in vivo imaging.

For Research Use Only. Not for use in diagnostic procedures.