missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Tyramide SuperBoost™ Kit, goat anti-rabbit IgG

Catalog No. p-7083200
Encompass
Change view
Click to view available options
Quantity:
50 Slides
Sufficient for 150 Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40943 50 Slides
B40922 Sufficient for 150 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40943 Supplier Invitrogen™ Supplier No. B40943
Only null left

Features poly-HRP-conjugated goat anti-rabbit IgG secondary antibody. Alexa Fluor™ 488 Tyramide SuperBoost™ Kit combine brightness of AlexaFluor dyes with high signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce s

SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).

Features:

  • High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
  • Simple protocol and detection using standard filters
  • Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
  • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments

Benefits:

  • Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
  • Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
  • Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
  • Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals

Shelf Life:

  • Stable for 6 months after receiving

Specifications

Quantity 50 Slides
Shipping Condition Approved for shipment at room temperature or on wet ice
Content And Storage 1 kit sufficient for 50 microscope slides (18 x 18 mm), containing:

• Blocking buffer (1X), 22.5 mL
• Poly-HRP-conjugated goat anti-rabbit secondary antibody (1X), 22.5 mL
• Alexa Fluor tyramide reagent
• Hydrogen peroxide (stabilized 3% solution), 28.5 mL
• Reaction buffer (20X), 6 mL
• Reaction stop solution, 8 mg
• Dimethylsulfoxide (DMSO), 200 μL

Store at 2-8°C. When stored as directed, the product is stable for 6 months after receiving.
Conjugate Alexa Fluor 488

For Research Use Only. Not for use in diagnostic procedures.