missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Hydrazide

Catalog No. a10436
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10436 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10436 Supplier Invitrogen™ Supplier No. A10436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor™ 488 Hydrazide is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins.

Alexa Fluor™ 488 Hydrazide is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins. Alexa Fluor™ 488 is a bright, green fluorescent dye with excitation ideally suited to the 488 nn laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 488 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 488 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

Detailed information about this AlexaFluor™ Hydrazide:
• Fluorophore label : Alexa Fluor™ 488 dye
• Reactive group: hydrazide
• Reactivity: Aldehydes or keytones in polysaccharides or glycoproteins
• Ex/Em of the conjugate: 493/517 nm
• Extinction coefficient: 71,000 cm-1M-1
• Spectrally similar dyes: FITC, GFP
• Molecular weight: 570.48

Cell Tracking and Tracing Applications
Alexa Fluor™ hydrazides and hydroxlamines are useful as low molecular weight, membrane-impermeant, aldehyde-fixable cell tracers, exhibiting brighter fluorescence and greater photostability than cell tracers derived from other spectrally similar fluorophores. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent.

Glycoprotein and Polysaccharide Labeling Applications
The Alexa Fluor™ hydrazides and hydroxlamines are reactive molecules that can be used to add a fluorescent label to biomolecules containing aldehydes or ketones. Aldehydes and ketones can be introduced into polysaccharides and glycoproteins by periodate-mediated oxidation of vicinal diols. Galactose oxidase can also be used to oxidize terminal galactose residues of glycoproteins to aldehydes.

Hydrazide vs Hydroxylamine
Hydrazine derivatives react with ketones and aldehydes to yield relatively stable hydrazones. Hydroxylamine derivatives (aminooxy compounds) react with aldehydes and ketones to yield oximes. Oximes are superior to hydrazones with respect to hydrolytic stability. Both hydrazones and oximes can be reduced with sodium borohydride (NaBH4) to further increase the stability of the linkage.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (Cat. No. D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (Cat. No. A33086)
Antibody Conjugate Purification Kit for 20-50 μg (Cat. No. A33087)
Antibody Conjugate Purification kit for 50-100 μg (Cat. No. A33088)

Specifications

Quantity 1 mg
Product Type Hydrazide
Content And Storage Store at room temperature and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Hydrazide
Label or Dye Alexa Fluor™ 488
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 517 nm
Excitation 493 nm
Show More Show Less
What dye can I use that is non-reactive and can show an injection site?

The non-reactive Alexa Fluor and Alexa Fluor hydrazide derivatives may be used for injection site visualization. Other options include the fluorescent polystyrene microspheres, FluoSpheres, and dye-conjugated dextrans. The hydrazide derivatives and 'fixable' dextrans are retained by cross-linking using an aldehyde-based fixative.

I injected a fluorescent tracer, but cannot detect it after tissue is fixed and sectioned. What am I doing wrong?

Confirm that the tracer you are using crosslinks to proteins or has a primary amine for fixation-either a hydrazide, lysine fixable dextran, or a protein conjugate.
Use aldehyde-based fixatives to cross link the amines on the tracer.
Inject a larger amount or higher concentration of the tracer. Tracers are generally injected at 1-20% concentrations (10 mg/mL or higher).
Confirm that you are using the correct fluorescent filter for detection. You can perform a spot test by pipetting a small amount of the undiluted stock solution of the tracer onto a slide, then view under the filter you are using on your microscope. This will confirm if the tracer fluorescence can be detected and the fluorescent microscope filter is working properly.
Review tissue fixation and handling procedures to confirm if any reagents or processing procedures could be affecting the tracer.

I have labeled my neurons with an Alexa Fluor conjugated biocytin to look at transport but I wanted to examine only retrograde transport and biocytin appears to be moving retrograde and anterograde. What should I do?

Observing both types of transport is typical for biocytin. The conjugated cholera toxin subunit B products have been observed to travel only retrogradely.

Do you have a neuronal tracer similar to Lucifer Yellow but in another fluorescent color?

Lucifer Yellow CH is a hydrazide, so any of our Alexa Fluor or fluorescent hydrazides could potentially be used. A listing of them can be found here. (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing/hydrazides-biocytins.html#prd)

How do I know which tracer to choose for my experiment?

Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:

Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)

What products do you have for neuronal tracing?

Please check out this web page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html) for details.


For Research Use Only. Not for use in diagnostic procedures.