missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 488 Goat Anti-Rabbit SFX Kit

Catalog No. A31627
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31627 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31627 Supplier Molecular Probes™ Supplier No. A31627

Alexa Fluor™ 488 Goat Anti-Rabbit SFX Kit

Alexa Fluor™ SFX Kits contain Image-iT™ FX signal enhancer (I36933) plus one of sixteen different Alexa Fluor™ dye-labeled secondary antibodies. These kits provide 400μg (0.2 mL of 2 mg/mL) of either goat anti-mouse IgG or goat anti-rabbit IgG antibody (as a standard or highly cross-adsorbed preparation) conjugated to Alexa Fluor™ 488, Alexa Fluor™ 555, Alexa Fluor™ 594, or Alexa Fluor™ 647 dye, four of our most commonly used Alexa Fluor™ dyes. The Alexa Fluor™ 488 goat anti-rabbit IgG included with this kit is also available in a 1000μg unit size (A11008).

Specifications

Conjugate Alexa Fluor 488
Description Alexa Fluor™ 488 Goat Anti-Rabbit SFX Kit, 1kit, Goat Host, Rabbit Species Reactivity, Alexa Fluor™ 488 Conjugate, Isotype, 2 to 8°C Storage Temperature, Room Temperature Shipping Condition
Host Species Goat
Quantity 1 Kit
Regulatory Status RUO
Whole Molecule Whole Antibody
Primary or Secondary Secondary
Packaging 1 Each
Target Species Rabbit
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Reactivity Rabbit
Shipping Condition Room Temperature
Product Line Alexa Fluor, Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.