missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Alkyne (Alexa Fluor™ 488 5-Carboxamido-(Propargyl), Bis(Triethylammonium Salt)), 5-isomer

Catalog No. A10267
Encompass
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10267 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10267 Supplier Invitrogen™ Supplier No. A10267

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor™ 488 Alkyne (Alexa Fluor™ 488 5-Carboxamido-(Propargyl), Bis(Triethylammonium Salt)), 5-isomer

The green-fluorescent Alexa Fluor™ 488 alkyne is reactive with azides via a copper-catalyzed click reaction. In addition to being an exceedingly bright and photostable fluorophore for use with flow cytometry, microscopy and HCS, Alexa Fluor™ 488 can also be utilized as a bio-orthogonal or biologically unique hapten for use in applications requiring signal amplification.

Specifications

Format Solid
Quantity 0.5 mg
Product Type Alkyne
Molecular Weight (g/mol) 773.91 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccate and protect from light.
Chemical Reactivity Azide
Detection Method Fluorescence
Reactive Group Alkyne
Shipping Condition Room Temperature
Reactive Moiety Alkyne
Label or Dye Alexa Fluor™ 488
Label Type Alexa Fluor
Color Green
Product Line Alexa Fluor, Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.