missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Alexa Fluor™ 420 NHS Ester (Succinimidyl Ester)
GREENER_CHOICE

Catalog No. A16302
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16302 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16302 Supplier Thermo Scientific™ Supplier No. A16302

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alexa Fluor 420 NHS Ester is an amine-reactive dye optimized for multiplex IHC and spatial biology. A long Stokes shift and 420/500 ex/em make it a good match for the CFP filter.

Alexa Fluor 420 NHS Ester is a fluorescence-based amine-reactive dye specifically optimized for multiplex immunohistochemistry and spatial biology. With excitation/emission maxima of 420/500 nm, this dye is a good match for the CFP filter on fluorescence microscopes.

Alexa Fluor dyes are reactive molecules that can be used to add a fluorescent label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting conjugates exhibit bright fluorescence and excellent photostability.

Typical conjugation reaction

Amine-reactive Alexa Fluor 420 NHS Ester can be conjugated with virtually any protein, antibody, or peptide. The reaction can be scaled for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

Alexa Fluor 420 NHS Ester is typically dissolved in high-quality, anhydrous dimethylsulfoxide (DMSO) (Cat. No. D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 15 minutes. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, more selective labeling of the amine terminus may be achieved using a buffer closer to neutral pH.

Conjugate purification

Labeled antibodies are typically separated from free Alexa Fluor dye using a gel filtration column, such as those included in the Antibody Labeling Flex Kit for 0.5-1 mg of antibody (Cat. No. A88072).

Specifications

For Use With (Application) Antibody and Protein Labeling
Quantity 100 μg
Product Type Amine Reactive Dye
Species Any
Molecular Weight (g/mol) ~700 g/mol
Solubility DMSO
Content And Storage -20°C
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument
Target Antibodies or Protein
Reactive Moiety NHS Ester
Label or Dye Alexa Fluor™ 420
Label Type Alexa Fluor
Color Green
Product Line Alexa Fluor
Emission 500 nm
Excitation 420 nm
Labeling Target Antibodies, Proteins
Excitation/Emission 420/500 nm
Show More Show Less

For Research Use Only.