missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alendronate Sodium Trihydrate 97.0+%, TCI America™
Supplier: TCI America A245625G
Specifications
| Alendronate Sodium Trihydrate | |
| White | |
| MFCD01748233 | |
| alendronate sodium hydrate, alendronate sodium trihydrate, alendronate sodium, alendronate monosodium trihydrate, alendros, adronat, fosamax, sodium alendronate hydrate, elandor, fosalan | |
| C(CC(O)(P(=O)(O)O)P(=O)(O)[O-])CN.O.O.O.[Na+] | |
| 325.122 | |
| CHEBI:2566 | |
| ≥97.0% (T) |
| 121268-17-5 | |
| C4H18NNaO10P2 | |
| 25 g | |
| DCSBSVSZJRSITC-UHFFFAOYSA-M | |
| sodium;(4-amino-1-hydroxy-1-phosphonobutyl)-hydroxyphosphinate;trihydrate | |
| 23681107 | |
| 271.08 | |
| Crystalline Powder |
Chemical Identifiers
| 121268-17-5 | |
| 325.122 | |
| DCSBSVSZJRSITC-UHFFFAOYSA-M | |
| 23681107 | |
| sodium;(4-amino-1-hydroxy-1-phosphonobutyl)-hydroxyphosphinate;trihydrate |
| C4H18NNaO10P2 | |
| MFCD01748233 | |
| alendronate sodium hydrate, alendronate sodium trihydrate, alendronate sodium, alendronate monosodium trihydrate, alendros, adronat, fosamax, sodium alendronate hydrate, elandor, fosalan | |
| CHEBI:2566 | |
| C(CC(O)(P(=O)(O)O)P(=O)(O)[O-])CN.O.O.O.[Na+] |
Safety and Handling
RTECSNumber : SZ6523666
TSCA : No
Recommended Storage : Freezer