missing translation for 'onlineSavingsMsg'
Learn More

Alendronate Sodium Trihydrate 97.0+%, TCI America™

Catalog No. p-7117921
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24565G 5 g
A245625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24565G Supplier TCI America Supplier No. A24565G
Only null left

Chemical Identifiers

CAS 121268-17-5
Molecular Formula C4H18NNaO10P2
Molecular Weight (g/mol) 325.122
MDL Number MFCD01748233
InChI Key DCSBSVSZJRSITC-UHFFFAOYSA-M
Synonym alendronate sodium hydrate, alendronate sodium trihydrate, alendronate sodium, alendronate monosodium trihydrate, alendros, adronat, fosamax, sodium alendronate hydrate, elandor, fosalan
PubChem CID 23681107
ChEBI CHEBI:2566
IUPAC Name sodium;(4-amino-1-hydroxy-1-phosphonobutyl)-hydroxyphosphinate;trihydrate
SMILES C(CC(O)(P(=O)(O)O)P(=O)(O)[O-])CN.O.O.O.[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 271.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Alendronate Sodium Trihydrate
RTECSNumber SZ6523666
TSCA No
Recommended Storage Freezer