Learn More
MilliporeSigma™ Supelco™ Aldrich™ Chromatography Sprayer
Provide a fine, uniform spray that is optimized for the development of TLC plates. Also suited for use in electrophoresis.
Supplier:  MilliporeSigma™ Supelco™ Z5297291EA
 View more versions of this product
Catalog No. 111009542
Only null left
Description
Description
Adjustable spray pattern using thumb on vent hole. Greaseless, screw-threaded joint will not seize; a simple turn of the threaded cap pulls joint apart safely. Threaded SafetyBarbs™ prevent accidental breakage during tubing installation or removal. Uses low-pressure gas or air.
Specifications
Chemical Identifiers
TLC
Aldrich
Specifications
50 mL
TLC
Aldrich
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC