Learn More
MilliporeSigma™ Supelco™ Aldrich™ Chromatography Sprayer
Catalog No. p-7191574
Change view
Click to view available options
Capacity (Metric):
10 mL
50 mL
125 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
111009541 10 mL
111009542 50 mL
111009543 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111009541 Supplier MilliporeSigma™ Supelco™ Supplier No. Z5297101EA
Only null left
Provide a fine, uniform spray that is optimized for the development of TLC plates. Also suited for use in electrophoresis.
Adjustable spray pattern using thumb on vent hole. Greaseless, screw-threaded joint will not seize; a simple turn of the threaded cap pulls joint apart safely. Threaded SafetyBarbs™ prevent accidental breakage during tubing installation or removal. Uses low-pressure gas or air.
Specifications
Capacity (Metric) 10 mL
For Use With (Application) TLC
Product Line Aldrich
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC