missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALDH16A1 Recombinant Rabbit Monoclonal Antibody (23GB4315)
GREENER_CHOICE

Catalog No. MA552682
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552682 100 μL
MA552683 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552682 Supplier Invitrogen™ Supplier No. MA552682
Only null left

Rabbit Recombinant Monoclonal Antibody

ALDH16A1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen ALDH16A1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4315
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ALDH16A1
Gene Accession No. Q3T1L0, Q571I9, Q8IZ83
Gene Alias 2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; aldehyde dehydrogenase family 16 member A1; ALDH16A1; LOW QUALITY PROTEIN: aldehyde dehydrogenase family 16 member A1; mFLJ00209
Gene Symbols ALDH16A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALDH16A1 (550-590AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126133, 361571, 69748
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less