missing translation for 'onlineSavingsMsg'
Learn More

Aldehyde-Ketone-DNPH TO-11A Calibration Mix, Restek

Catalog No. 08776252
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08776252 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08776252 Supplier Chromatographic Supplier No. RK 31808

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Analyte or Component Names 15 components:
acetaldehyde-DNPH,
acetone-DNPH,
acrolein-DNPH
Concentration 15 μg/mL
Element Acetaldehyde-DNPH, Acetone-DNPH, Acrolein-DNPH
Solvent or Matrix Acetonitrile
Quantity 1 mL
Recommended Storage Store in a cool dry ventilated location. Keep container closed away from sources of ignition.