missing translation for 'onlineSavingsMsg'
Learn More

Alcohol (HPLC, Meets ACS), Fisher Chemical™

$390.25 - $390.25

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A995-4 Edge
SDS Accessories View Documents
Thermo Fisher Scientific
BPA9954
4 L Glass Bottle
Each for $390.25
Case of 4 Each for $1,561.00
Only null left
Edge
 
Description

Description

Specifications

Chemical Identifiers

64-17-5
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
702
Ethanol
C2H6O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:16236
CO

Specifications

-114.1°C
0.04 AU max. at 254nm, 0.35 AU max. at 220nm, 0.65 AU max. at 210nm, 1 AU max. at 205nm
78.5°C
Pass Test
Liquid
5ppm max.
0.0003mEq/g max.
0.0002mEq/g max.
MFCD00004595
1987; Clear
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:16236
Anhydrous Ethyl Alcohol 90% ±1% v/v; Methyl Alcohol approx. 5% v/v; 2-Propanol approx. 5% v/v
64-17-5,67-56-1,67-63-0
1.24 (Air= 1.0)
4 L
Glass Bottle
1.3580 to 1.3610 (at 25°C)
99.8% min by GC
C2H6O
CH3CH2OH
44mmHg
0.1% max.
OKKJLVBELUTLKV-UHFFFAOYSA-N
Ethanol
702
Mobile phase for HPLC applications
Alcohol
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Harmful by inhalation, in contact with skin and if swallowed. Vapor harmful. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment.

NFPA

Health:2
Flammability:3
Instability:0

SDS
missing translation for 'documents'

missing translation for 'documents'