Learn More
Thermo Scientific Chemicals Alcohol dehydrogenase, yeast
Catalog No. AAJ65869WEX
Change view
Click to view available options
Quantity:
150 kilounits
30 kilounit
75 kilounits
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65869WEX 150 kilounits
AAJ658699PL 30 kilounit
AAJ65869VEX 75 kilounits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ65869WEX Supplier Thermo Scientific Chemicals Supplier No. J65869WEX
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alcohol dehydrogenase catalyzes the reaction: RCH2OH +NAD+ ↔ RCHO + NADH + H+ It facilitates the interconversion between alcohols and aldehydes or ketones with the reduction of nicotinamide adenine dinucleotide (NAD+ to NADH). In biotransformation, alcohol dehydrogenases are often used for the synthesis of enantiomerically pure stereoisomers of chiral alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity 150 kilounits
Packaging Glass bottle
Form Lyophilized
RUO – Research Use Only