Learn More
Thermo Scientific Chemicals Alcohol dehydrogenase, yeast
Supplier: Thermo Scientific Chemicals J65869VEX
Description
Alcohol dehydrogenase catalyzes the reaction: RCH2OH +NAD+ ↔ RCHO + NADH + H+ It facilitates the interconversion between alcohols and aldehydes or ketones with the reduction of nicotinamide adenine dinucleotide (NAD+ to NADH). In biotransformation, alcohol dehydrogenases are often used for the synthesis of enantiomerically pure stereoisomers of chiral alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| Glass bottle | |
| Lyophilized |
Specifications
| 75 kilounits | |
| Glass bottle | |
| Lyophilized |
RUO – Research Use Only