missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alcohol Dehydrogenase 1A Recombinant Rabbit Monoclonal Antibody (23GB4295)

Catalog No. MA552679
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552679 20 μL
MA552678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552679 Supplier Invitrogen™ Supplier No. MA552679
Only null left

Rabbit Recombinant Monoclonal Antibody

Alcohol Dehydrogenase 1A Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of the alcohol dehydrogenase family. The encoded protein is the alpha subunit of class I alcohol dehydrogenase, which consists of several homo- and heterodimers of alpha, beta and gamma subunits. Alcohol dehydrogenases catalyze the oxidation of alcohols to aldehydes. This gene is active in the liver in early fetal life but only weakly active in adult liver. This gene is found in a cluster with six additional alcohol dehydrogenase genes, including those encoding the beta and gamma subunits, on the long arm of chromosome 4. Mutations in this gene may contribute to variation in certain personality traits and substance dependence.

Specifications

Antigen Alcohol Dehydrogenase 1A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4295
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADH1A
Gene Accession No. P00329, P06757, P07327
Gene Alias ADC1; Adh; ADH, alpha subunit; ADH1; Adh-1; ADH1A; Adh1c; Adh-1e; Adh1-e; Adh-1t; Adh-1-t; Adh1-t; Adh1tl; Adh-3e; Adh3-e; ADH-A2; ADH-AA; AI194826; Alcohol dehydrogenase 1; alcohol dehydrogenase 1 (class I); alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1 temporal, liver; alcohol dehydrogenase 1, complex; alcohol dehydrogenase 1, electrophoretic; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 3, electrophoretic; alcohol dehydrogenase A subunit; alcohol dehydrogenase ADH1; Alcohol dehydrogenase I; Alcohol dehydrogenase subunit alpha; aldehyde reductase; class I alcohol dehydrogenase; O0947; YADH-1; YOL086C
Gene Symbols Adh1, ADH1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human Alcohol Dehydrogenase (1-50AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11522, 124, 24172
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less