missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alcian Blue Solution, For Microscopy, MilliporeSigma™
Specifications
Specifications
| Odor | Weak smell of acetic acid |
| Packaging | Plastic bottle |
| Color | Dark Blue |
| pH | 2.5 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Grade | Not Available |
| Chemical Name or Material | Alcian Blue Solution |