missing translation for 'onlineSavingsMsg'
Learn More

Albuterol Related Compound B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014164
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014164 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014164 Supplier Merck Emd Millipore Supplier No. PHR204050MG
Only null left

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 41489-89-8
Linear Formula C13H19 NO3 · HCl
Synonym 2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone hydrochloride
Type Certified Reference Material
Quantity 50 mg
Formula Weight 273.76
Grade Certified Reference Material