missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Albumin, Bovine, Fraction V Powder

Catalog No. mp08810025
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08810025 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08810025 Supplier MP Biomedicals™ Supplier No. 08810025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Albumin is used to solubilize lipids, and is also used as a blocking agent in Western blots or ELISA applications.

Cream, crystalline “flaky” powder. Prior to lyophilization, the product is filtered through a 0.2mm filter. Low ionic strength and bioburden.

  • Solubility: 0.042
  • pH: 7.3 (7% solution in 0.15M NaCl)
  • Purity: 100%
  • Loss on Drying: 3.5%
  • Heavy Metals: <10ppm
  • Additional Tests: Vesicular Stomatitis Virus: Negative; Bluetongue Virus: Negative
  • General: This product was derived from bovine blood collected at a USDA licensed facility. The cattle received ante- and post-mortem health inspection under a veterinarian's supervision and were apparently free from infectious and contagious diseases. All donor animals were sourced from the United States. Veterinary Certificates are maintained on file at the manufacturing site.

Specifications

For Use With (Application) Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol) ∽66
Name Bovine Blood Albumin Fraction V
pH Range 6.8 to 7.2
Preparation Method Prior to lyophilization, the product is filtered through a 0.2μm filter
Quantity 1 kg
Source Bovine Blood
Storage Requirements 2° to 8°C
Form Powder