missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Alanine methyl ester hydrochloride, 98%
CAS: 3196-73-4 | C4H10ClNO2 | 139.58 g/mol
$401.78
Chemical Identifiers
| CAS | 3196-73-4 |
|---|---|
| Molecular Formula | C4H10ClNO2 |
| Molecular Weight (g/mol) | 139.58 |
| MDL Number | MFCD00039060 |
| InChI Key | XPGRZDJXVKFLHQ-UHFFFAOYSA-N |
| Synonym | methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride |
| PubChem CID | 2734767 |
| IUPAC Name | methyl 3-aminopropanoate;hydrochloride |
| SMILES | Cl.COC(=O)CCN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC371470250
|
N/A | 25g, 25 g | Glass bottle |
Each for $401.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C4H10ClNO2 | |
| MFCD00039060 | |
| methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride | |
| methyl 3-aminopropanoate;hydrochloride |
Specifications
| 103.0°C to 105.0°C | |
| 97.5 | |
| White | |
| 98% | |
| NH2CH2CH2COOCH3·HCl | |
| 04, IV, 2527 | |
| XPGRZDJXVKFLHQ-UHFFFAOYSA-N | |
| methyl 3-aminopropanoate;hydrochloride | |
| 2734767 | |
| 98% | |
| β-Alanine methyl ester hydrochloride, 98% |
| 3196-73-4 | |
| 100.0 | |
| Authentic | |
| C4H10ClNO2 | |
| MFCD00039060 | |
| methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride | |
| Cl.COC(=O)CCN | |
| 139.58 | |
| 139.58 | |
| Crystalline Powder |
RUO – Research Use Only