missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine methyl ester hydrochloride, 98%

Catalog No. AC371470250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC371470250 25 g Glass Bottle
AC371470050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC371470250 Supplier Thermo Scientific Chemicals Supplier No. 371470250

CAS: 3196-73-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3196-73-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00039060
InChI Key XPGRZDJXVKFLHQ-UHFFFAOYSA-N
Synonym methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
PubChem CID 2734767
IUPAC Name methyl 3-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)CCN

Specifications

Melting Point 103.0°C to 105.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula NH2CH2CH2COOCH3·HCl
Quantity 25 g
Beilstein 04, IV, 2527
Formula Weight 139.58
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material β-Alanine methyl ester hydrochloride, 98%
Show More Show Less

RUO – Research Use Only