missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals β-Alanine, 98%

Catalog No. p-7023273
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1666530 250 g
AAA166650B 1000 g
AAA166650I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1666530 Supplier Thermo Scientific Chemicals Supplier No. A1666530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-95-9 | C3H7NO2 | 89.09 g/mol

Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.

Solubility
Soluble in water(550g/L). Slightly soluble in alcohol. Insoluble in ether and acetone.

Notes
Slightly hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 107-95-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008200
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
PubChem CID 239
ChEBI CHEBI:16958
IUPAC Name 3-aminopropanoic acid
SMILES NCCC(O)=O

Specifications

Melting Point ∽205°C (decomposition)
Assay Percent Range 98%
Linear Formula H2NCH2CH2CO2H
Beilstein 906793
Merck Index 14,205
Solubility Information Soluble in water(550g/L). Slightly soluble in alcohol. Insoluble in ether and acetone.
Density 1.437 g/mL
Quantity 250 g
Formula Weight 89.09
Percent Purity 98%
Chemical Name or Material β-Alanine
Show More Show Less
EINECSNumber 203-536-5
RTECSNumber UA2369200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only