missing translation for 'onlineSavingsMsg'
Learn More

Alachlor, SPEX CertiPrep™

Catalog No. S185
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S185 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S185 Supplier SPEX CertiPrep Group LLC Supplier No. S185
Only null left

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Alachlor15972-60-81000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 15972-60-8 , 67-56-1
Molecular Formula C14H20ClNO2
Molecular Weight (g/mol) 269.769
InChI Key XCSGPAVHZFQHGE-UHFFFAOYSA-N
PubChem CID 2078
ChEBI CHEBI:2533
IUPAC Name 2-chloro-N-(2,6-diethylphenyl)-N-(methoxymethyl)acetamide
SMILES CCC1=C(C(=CC=C1)CC)N(COC)C(=O)CCl

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Methods For US EPA Method 8081
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol
Quantity 1 mL
Chemical Name or Material Alachlor
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only