missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKR1B1 Monoclonal Antibody (K92029_3F2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555628
Description
Sequence of this protein is as follows: AKMPILGLGT WKSPPGQVTE AVKVAIDVGY RHIDCAHVYQ NENEVGVAIQ EKLREQVVKR EELFIVSKLW CTYHEKGLVK GACQKTLSDL KLDYLDLYLI HWPTGFKPGK EFFPLDESGN VVPSDTNILD TWAAMEELVD EGLVKAIGIS NFNHLQVEMI LNKPGLKYKP AVNQIECHPY LTQEKLIQYC QSKGIVVTAY SPLGSPDRPW AKPEDPSLLE DPRIKAIAAK HNKTTAQVLI RFPMQRNLVV IPKSVTPERI AENFKVFDFE LSSQDMTTLL
Catalyzes the NADPH-dependent reduction of a wide variety of carbonyl-containing compounds to their corresponding alcohols with a broad range of catalytic efficiencies.Specifications
| AKR1B1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P07943, P15121, P45376 | |
| AKR1B1, Akr1b3 | |
| Recombinant human AKR1B1 (immunogen range is 11-290). | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| K92029_3F2 | |
| Unconjugated | |
| AKR1B1 | |
| ADR; Ahr1; Ahr-1; Akr1b1; Akr1b3; Akr1b4; Aldehyde reductase; aldehyde reductase 1; Aldehyde reductase 1 (low Km aldose reductase) (5.8 kb PstI fragment, probably the functional gene); aldo-keto reductase family 1; aldo-keto reductase family 1 member B; aldo-keto reductase family 1 member B1; aldo-keto reductase family 1 member B1; aldose reductase; Aldo-keto reductase family 1 member B3; aldo-keto reductase family 1, member B1 (aldose reductase); aldo-keto reductase family 1, member B3 (aldose reductase); aldo-keto reductase family 1, member B4 (aldose reductase); Aldor1; Aldose reductase; ALDR1; ALDRED; Alr; Alr2; ALR-P-I; AR; Lii5-2 CTCL tumor antigen; low Km aldose reductase; member B1 (aldose reductase); RATALDRED | |
| Mouse | |
| Protein A | |
| RUO | |
| 11677, 231, 24192 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |