missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKNA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565717
Description
Immunogen sequence: LDMTEEEPDGT LGSLEVEEAG ESSSRLGYEA GLSLEGHGNT SPMALGHGQA RGWVASGEQA SGDKLSEHSE VNPSVELSPA RSWSSGTVSL DHPSDSLDST WEG Highest antigen sequence identity to the following orthologs - mouse 53%, rat 51%.
AKNA is a transcription factor that specifically activates the expression of the CD40 receptor and its ligand CD40L/CD154, two cell surface molecules on lymphocytes that are critical for antigen-dependent-B-cell development. It binds to A/T-rich promoters.Specifications
| AKNA | |
| Polyclonal | |
| Unconjugated | |
| AKNA | |
| AKNA; AKNA transcript F2; AT-hook transcription factor; AT-hook transcription factor AKNA; AT-hook-containing transcription factor; KIAA1968; Microtubule organization protein AKNA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80709 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z591 | |
| AKNA | |
| Recombinant Human AKNA. Recombinant protein control fragment (Product #RP-106388). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |