missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523602

Catalog No. PIPA523602

Only null left
Explore available promotions

Description

Description

AKIP1 Polyclonal Antibody for Western Blot

Enhances NF-kappa-B transcriptional activity by regulating the nuclear localization of the NF-kappa-B subunit RELA and promoting the phosphorylation of RELA by PRKACA. Regulates the effect of the cAMP-dependent protein kinase signaling pathway on the NF-kappa-B activation cascade.
Specifications

Specifications

AKIP1
Polyclonal
Unconjugated
Akip1
A kinase (PRKA) interacting protein 1; A-kinase interacting protein 1; A-kinase-interacting protein 1; Akip1; BCA3; breast cancer associated 3; breast cancer associated gene 3; breast cancer-associated gene 3 protein; C11orf17; D7H11orf17; D930014E17Rik; ICRFP703B1614Q5.6; ICRFP703N2430Q5.6; koyt binding protein 1; koyt binding protein 2; koyt binding protein 3; ORF27; PKA-interacting protein; Proline-rich protein BCA3; protein kinase A-interacting protein 1; RGD1306959
Rabbit
Antigen affinity chromatography, Protein A
RUO
100751192, 56672, 57373
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9JJR5, Q9NQ31
Akip1
KLH conjugated synthetic peptide between 54-83 amino acids from the N-terminal region of human BCA3.
400 μL
Primary
Human, Mouse, Hamster
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions