missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKAP8 Recombinant Rabbit Monoclonal Antibody (23GB2970)
GREENER_CHOICE

Catalog No. MA553326
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553326 100 μL
MA553327 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553326 Supplier Invitrogen™ Supplier No. MA553326
Only null left

Rabbit Recombinant Monoclonal Antibody

AKAP8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The type II cAMP-protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is thought to be mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins. The family of RII-anchoring proteins has been designated A-kinase anchoring proteins (AKAP). AKAP 95, also known as AKAP 8, is a nuclear matrix protein predominantly expressed in liver, heart, pancreas, kidney and skeletal muscle. During mitosis, AKAP 95 is recruited to the chromosomes and plays an essential role in mitotic progression. Characteristic of its family, AKAP 95 participates in PKA signaling through an interaction with the RII regulatory subunit. In addition, AKAP 95 forms a complex with HA95 and HDAC3 and is required for the deacetylation of Histone H3 in mitosis.

Specifications

Antigen AKAP8
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2970
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKAP8
Gene Accession No. O43823, Q63014, Q9DBR0
Gene Alias 1200016A02Rik; A kinase (PRKA) anchor protein 8; A kinase anchor protein 8; AA673585; AKAP 8; AKAP 95; Akap8; AKAP-8; Akap95; AKAP-95; A-kinase anchor protein 8; A-kinase anchor protein 95 kDa; A-kinase anchor protein, 95kDa; A-kinase anchoring protein 8; AU015639; DKFZp586B1222
Gene Symbols AKAP8
Host Species Rabbit
Immunogen A synthesized peptide derived from AKAP 95 (30-55AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10270, 116633, 56399
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less