missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKAP14 Recombinant Rabbit Monoclonal Antibody (23GB3935)

Catalog No. MA552643
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552643 20 μL
MA552642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552643 Supplier Invitrogen™ Supplier No. MA552643
Only null left

Rabbit Recombinant Monoclonal Antibody

AKAP14 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The protein anchors PKA in ciliary axonemes and, in this way, may play a role in regulating ciliary beat frequency. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specifications

Antigen AKAP14
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3935
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AKAP14
Gene Accession No. O35817, Q86UN6
Gene Alias A kinase (PRKA) anchor protein 14; AKAP 28; AKAP14; AKAP-14; AKAP28; A-kinase anchor protein 14; A-kinase anchor protein 28 kDa; A-kinase anchoring protein 14; A-kinase anchoring protein 28; PRKA14; Protein kinase A-anchoring protein 14; TAKAP-1.2; Takap80; Testis-specific A-kinase-anchoring protein TAKAP-80; Testis-specific A-kinase-anchoring-protein
Gene Symbols AKAP14
Host Species Rabbit
Immunogen A synthesized peptide derived from human AKAP14 (100-167AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158798, 434756, 60332
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less