missing translation for 'onlineSavingsMsg'
Learn More

AK Bouffant/Bootie Dispenser

Dispense booties, bouffants, and other similarly sized apparel with ease. The AK Bouffant/Bootie Dispenser features a round, front-based center opening for easy removal of stored apparel and is available in two sizes.

Supplier:  AK AK262

 View more versions of this product

Catalog No. 17-987-140K


Only null left
Explore available promotions

Description

Description

  • Versatile design allows for dispensation of a variety of apparel supplies
  • Top-loading hinged lid for easy re-stocking
  • Can be wall-mounted or used free standing
  • Made of 0.125 in high impact PETG
Specifications

Specifications

Apparel Dispenser
10 in.
10 x 10 x 10 in.
25.4 cm
25.4 cm
Free-standing or Wall Mount
0.125 in.
Clear
25.4 cm
10 in.
10 in.
PETG
Hinged Lid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions