missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AIRE Monoclonal Antibody (5H12), eFluor™ 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Invitrogen™ 50593482

 View more versions of this product

Catalog No. 501124413


Only null left
Explore available promotions

Description

Description

Description: This 5H12 monoclonal antibody reacts with mouse Autoimmune Regulator (AIRE). This 58-kDa transcription factor is highly expressed within the perinuclear region of mature medullary thymic epithelial cells (mTECs). However, expression of AIRE protein in the periphery remains unclear despite the detection of AIRE mRNA in the lymph node, lymphoid stromal cells, monocytes, and dendritic cells. Studies demonstrate that AIRE is involved in thymocyte development, in particular the regulation of tissue-specific antigen expression during negative selection. Other biological functions for AIRE include chemokine expression and antigen presentation. The 5H12 antibody recognizes mouse AIRE isoforms 1 and 2. Applications Reported: This 5H12 antibody has been reported for use in intracellular flow cytometric analysis and immunohistochemical staining of frozen tissue sections. Applications Tested: This 5H12 antibody has been tested by immunohistochemistry on frozen 5-week-old mouse thymus and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

AIRE is a transcriptional regulator protein that binds to DNA as a dimer and tetramer, but not as a monomer, and may be involved in the regulation of gene expression. A nuclear protein, AIRE is a 545 amino acid protein with an HSR domain, two PHD type zinc fingers, and a SAND domain, and may be present in soluble high-molecular-weight complexes. It may form homodimer, homotetramer and also nuclear bodies and interacts with the transcriptional coactivator CBP and CREBBP. Three isoforms of AIRE are reported and one of them is predicted to be a candidate for nuclear-mediated decay (NMD). AIRE is involved in immune regulation and is known to mediate E3 ubiquitin ligase activity. Defects in AIRE are responsible for the rare autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED).
Specifications

Specifications

AIRE
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9Z0E3
Aire
Affinity chromatography
RUO
11634
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
5H12
eFluor 660
Aire
Aire; AIRE protein; AIRE1; APECED; APECED protein; APECED protein homolog; APS1; APSI; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein homolog; Autoimmune regulator; autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy); autoimmune regulator AIRE1a; LOW QUALITY PROTEIN: autoimmune regulator; OTTHUMP00000109529; PGA1
Rat
100 μg
Primary
Mouse
Antibody
IgG2c κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.