missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ AIN-93M Rodent Diet
Purified diet formulated to meet the nutritional requirements of mature rodents
$84.71
Specifications
| Product Type | Rodent Diet |
|---|---|
| Components | Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ |
| For Use With (Application) | Used and published for research objectives |
| Species | Rodent |
Description
- Widely used and published
- Phytoestrogen free
- Chlorophyll free—does not interfere with imaging studies
- Nutritionally assayed
- Ingredients: Corn Starch, Casein, Maltodextrin, Sucrose, Cellulose, Soybean Oil, Mineral Mix, Vitamin Mix, Choline Bitartrate, L-Cystine, tBHQ
Specifications
| Rodent Diet | |
| Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ | |
| Used and published for research objectives | |
| Rodent |
Safety and Handling
ShelfLife : 6 Months