missing translation for 'onlineSavingsMsg'
Learn More

Germaine™ Laboratories AimStrip™ Dipstick Reagent Strips

Germaine™ Laboratories AimStrip™ Dipstick Reagent Strips are for qualitative detection of analytes in urine

$24.10

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Sample Type Urine
Type Urinalysis Dipstick
Disposable Single-use
View More Specs

Products 1
Catalog Number Mfr. No. Detectable Analytes Sensitivity Price Quantity & Availability  
Catalog Number Mfr. No. Detectable Analytes Sensitivity Price Quantity & Availability  
23111291
View Documents Promotion Details
Germaine™ Laboratories
50121
Ketone 5 to 10 mg/dL Acetoacetic Acid
Pack of 100 Each for $24.10
Case of 12 Pack for $289.20
Only null left
 
Description

Description

Specifications

Specifications

2°C to 30°C
Urine
Single-use
100 Tests
Waived
Urinalysis Dipstick
Urinalysis
Videos
SDS
Documents

Documents