missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germaine™ Laboratories AimStrip™ Dipstick Reagent Strips
Specifications
Specifications
| Content And Storage | 2°C to 30°C |
| Clia Complexity | Waived |
| Detectable Analytes | Ketone |
| Sample Type | Urine |
| Sensitivity | 5 to 10 mg/dL Acetoacetic Acid |
| Type | Urinalysis Dipstick |
| Disposable | Single-use |
| For Use With (Application) | Urinalysis |
| Quantity | 100 Tests |