missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AHA1 Recombinant Rabbit Monoclonal Antibody (23GB3170)
GREENER_CHOICE

Catalog No. p-200056705
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552600 100 μL
MA552601 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552600 Supplier Invitrogen™ Supplier No. MA552600
Only null left

Rabbit Recombinant Monoclonal Antibody

AHA1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Acts as a co-chaperone of HSP90AA1 (PubMed:29127155). Activates the ATPase activity of HSP90AA1 leading to increase in its chaperone activity (PubMed:29127155). Competes with the inhibitory co-chaperone FNIP1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:27353360). Competes with the inhibitory co-chaperone TSC1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:29127155). [UniProt].

Specifications

Antigen AHA1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3170
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ahsa1
Gene Accession No. O95433, Q8BK64
Gene Alias Activator of 90 kDa heat shock protein ATPase homolog 1; activator of Hsp90 ATPase activity 1; Activator of Hsp90 ATPase protein 1; Aha 1; AHA1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast); AHA1, activator of heat shock protein ATPase 1; AHA1, activator of heat shock protein ATPase homolog 1; Aha1p; AHSA 1; Ahsa1; BC023857; C14orf3; hAha1; Hsp90 co-chaperone AHA1; HSPC322; p38; SHSA1; YD8142.16; YD8142B.06; YDR214W
Gene Symbols Ahsa1
Host Species Rabbit
Immunogen A synthetic peptide of human AHA1 (300-338AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10598, 217737, 681996
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less