missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AGL Recombinant Rabbit Monoclonal Antibody (23GB2965)
GREENER_CHOICE

Catalog No. MA553592
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553592 100 μL
MA553593 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553592 Supplier Invitrogen™ Supplier No. MA553592
Only null left

Rabbit Recombinant Monoclonal Antibody

AGL Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described.

Specifications

Antigen AGL
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2965
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AGL
Gene Accession No. P35573
Gene Alias 1-Apr; 4-alpha-glucanotransferase; 4-glucantransferase; AGL; amylo-1, 6-glucosidase, 4-alpha-glucanotransferase; Amylo-1,6-glucosidase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase; amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferaseprovided; Amylo-alpha-1,6-glucosidase; Dextrin 6-alpha-D-glucosidase; GDE; glycogen debrancher; Glycogen debranching enzyme; glycogen debranching protein; Oligo-1; Oligo-1,4-1,4-glucantransferase
Gene Symbols AGL
Host Species Rabbit
Immunogen A synthesized peptide derived from AGL (60-90AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 178, 362029, 77559
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less